2,4,6-Trichloro-5-isopropylpyrimidine 95%

Supplier: Ambeed
Warning

A788005-100MG A788005-1G A788005-250MG
76993-210EA 32 USD
76993-210 76993-212 76993-214
2,4,6-Trichloro-5-isopropylpyrimidine 95%
2,4,6-Trichloro-5-isopropylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₇Cl₃N₂
MDL Number: MFCD19441365
CAS Number: 1780-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR