(R)-(+)-1-[2-(Diphenylphosphino)phenyl]ethylamine ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (R)-1-(2-(Diphenylphosphino)phenyl)ethanamine

A787520-1G A787520-250MG A787520-100MG
76993-206EA 336 USD
76993-206 76993-208 77886-760
(R)-(+)-1-[2-(Diphenylphosphino)phenyl]ethylamine ≥98%, ee 99%
(R)-(+)-1-[2-(Diphenylphosphino)phenyl]ethylamine

Precaución: for Research Use Only

Formula: C₂₀H₂₀NP
MDL Number: MFCD17013986
CAS Number: 192057-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR