2,2'-(1,2-Phenylenebis(oxy))diethanamine 95%

Supplier: Ambeed
Warning

A786998-100MG A786998-1G A786998-250MG
77019-298EA 111.4 USD
77019-298 77019-300 77019-302
2,2'-(1,2-Phenylenebis(oxy))diethanamine 95%
2,2'-(1,2-Phenylenebis(oxy))diethanamine

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₂
MW: 196.24 g/mol
MDL Number: MFCD00464521
CAS Number: 42988-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR