2-Quinolin-2-yl-1-(4-trifluoromethylphenyl)ethanol 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Quinolin-2-yl)-1-(4-(trifluoromethyl)phenyl)ethanol

A786808-1G A786808-250MG
77006-466EA 21 USD
77006-466 77006-468
2-Quinolin-2-yl-1-(4-trifluoromethylphenyl)ethanol 97%
2-Quinolin-2-yl-1-(4-trifluoromethylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₈H₁₄F₃NO
MDL Number: MFCD03424493
CAS Number: 496947-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR