(2S)-2-(4-(3-Amino-1,9-dioxo-1,2,5,6,6a,7-hexahydroimidazo[1,5-f]pteridin-8(9H)-yl)benzamido)pentanedioic acid 95%
Supplier: Ambeed
|
Warning
|
A786224-10MG
A786224-1MG
A786224-25MG
A786224-50MG
A786224-5MG
77019-282EA
222.99
USD
77019-282
77019-284
77019-286
77019-288
77019-290
(2S)-2-(4-(3-Amino-1,9-dioxo-1,2,5,6,6a,7-hexahydroimidazo[1,5-f]pteridin-8(9H)-yl)benzamido)pentanedioic acid 95%
(2S)-2-(4-(3-Amino-1,9-dioxo-1,2,5,6,6a,7-hexahydroimidazo[1,5-f]pteridin-8(9H)-yl)benzamido)pentanedioic acid
Precaución: for Research Use Only
Formula:
C₂₀H₂₁N₇O₇ |
MDL Number:
MFCD29920315 CAS Number: 10538-99-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...