Methyl 4-(2-hydroxyethyl)benzoate 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A785984-100MG A785984-1G A785984-250MG A785984-5G
76993-200EA 13 USD
76993-200 76993-202 76993-204 77684-726
Methyl 4-(2-hydroxyethyl)benzoate 95%, Ambeed.Inc
Methyl 4-(2-hydroxyethyl)benzoate

Precaución: For research use only.

Formula: C₁₀H₁₂O₃
MDL Number: MFCD24447574
CAS Number: 46190-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR