1-(Trifluoromethyl)cyclopropanecarboxylic acid 98%

Supplier: Ambeed
Danger

Synonyms: 1-(Trifluoromethyl)cyclopropane-1-carboxylic acid

A785405-1G A785405-5G A785405-25G A785405-10G
77019-274EA 18.04 USD
77019-274 77019-276 77552-432 77595-246
1-(Trifluoromethyl)cyclopropanecarboxylic acid 98%
1-(Trifluoromethyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅F₃O₂
MW: 154.09 g/mol
MDL Number: MFCD03093070
CAS Number: 277756-46-4
UN: 2923
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR