Formamidine disulfide dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Dithiourea Dihydrochloride, Dithiodiformamide Dihydrochloride

A785146-100G A785146-25G A785146-500G
77006-458EA 40 USD
77006-458 77006-460 77006-462
Formamidine disulfide dihydrochloride 98%
Formamidine disulfide dihydrochloride

Precaución: for Research Use Only

Formula: C₂H₆N₄S₂
MW: 223.13 g/mol
MDL Number: MFCD00012586
CAS Number: 14807-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR