Decaethylene glycol monomethyl ether 95%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
mPEG10-Alcohol, m-PEG10-alcohol, 2,5,8,11,14,17,20,23,26,29-Decaoxahentriacontan-31-ol
A784100-100MG
A784100-1G
A784100-250MG
A784100-5G
A784100-25G
77019-266EA
10
USD
77019-266
77019-268
77019-270
77684-725
77891-532
Decaethylene glycol monomethyl ether 95%, Ambeed.Inc
Decaethylene glycol monomethyl ether
Precaución: For research use only.
Formula:
C₂₁H₄₄O₁₁ MW: 472.57 g/mol |
MDL Number:
MFCD06797087 CAS Number: 27425-92-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...