1-(4-Methoxyphenyl)-3-methylbutan-2-one 95%

Supplier: Ambeed
Warning

A783227-100MG A783227-1G A783227-250MG
77019-252EA 55.17 USD
77019-252 77019-254 77019-256
1-(4-Methoxyphenyl)-3-methylbutan-2-one 95%
1-(4-Methoxyphenyl)-3-methylbutan-2-one

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
MW: 192.25 g/mol
MDL Number: MFCD11935284
CAS Number: 61173-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR