Methyl 4-Bromo-3-chlorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-3-chlorobenzoic acid methyl ester, Methyl 4-bromo-3-chlorobenzoate

A783135-100G A783135-1G A783135-25G A783135-5G
77019-244EA 790.38 USD
77019-244 77019-246 77019-248 77019-250
Methyl 4-Bromo-3-chlorobenzoate 98%
Methyl 4-Bromo-3-chlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MW: 249.49 g/mol
MDL Number: MFCD11045619
CAS Number: 117738-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR