3-Bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine 98%

Supplier: Ambeed
Warning

A783023-1G A783023-250MG A783023-5G
77006-448EA 42 USD
77006-448 77006-450 77006-452
3-Bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine 98%
3-Bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₈H₄BrF₃N₂
MW: 265.04 g/mol
MDL Number: MFCD12827771
CAS Number: 1146615-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR