2-Iodo-4-methoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A782721-100MG A782721-250MG
76993-192EA 65 USD
76993-192 76993-194
2-Iodo-4-methoxybenzaldehyde 95%
2-Iodo-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇IO₂
MDL Number: MFCD18452131
CAS Number: 105469-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR