2-Fluoro-1,3-dimethylimidazolidinium hexafluorophosphate 95%

Supplier: Ambeed
Warning

A782641-10G A782641-1G A782641-25G A782641-5G
77006-440EA 48 USD
77006-440 77006-442 77006-444 77006-446
2-Fluoro-1,3-dimethylimidazolidinium hexafluorophosphate 95%
2-Fluoro-1,3-dimethylimidazolidinium hexafluorophosphate

Precaución: for Research Use Only

Formula: C₅H₁₀FN₂·F₆P
MDL Number: MFCD01863767
CAS Number: 164298-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR