5-Norbornene-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Bicyclo[2.2.1]hept-5-ene-2-carboxylic Acid

A782160-100G A782160-10G A782160-25G A782160-500G A782160-5G
77006-430EA 33.95 USD
77006-430 77006-432 77006-434 77006-436 77006-438
5-Norbornene-2-carboxylic acid 97%
5-Norbornene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
MDL Number: MFCD00085356
CAS Number: 120-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR