Benzyl Spiro[indoline-3,4'-piperidine]-1'-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 1'-Benzyloxycarbonyl-spiro[indoline-3,4'-piperidine], 1'-Cbz-spiro[indoline-3,4'-piperidine], Spiro[indoline-3,4'-piperidine]-1'-carboxylic acid Benzyl ester, Benzyl spiro[indoline-3,4'-piperidine]-1'-carboxylate

A782114-100MG A782114-250MG A782114-5G
77019-238EA 27.59 USD
77019-238 77019-240 77019-242
Benzyl Spiro[indoline-3,4'-piperidine]-1'-carboxylate 97%
Benzyl Spiro[indoline-3,4'-piperidine]-1'-carboxylate

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₂O₂
MW: 322.41 g/mol
MDL Number: MFCD06738866
CAS Number: 167484-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR