4-Bromo-N-cyclopropyl-2-fluorobenzamide 98%

Supplier: Ambeed
Warning

A781790-1G A781790-250MG A781790-5G
77019-226EA 41.99 USD
77019-226 77019-228 77019-230
4-Bromo-N-cyclopropyl-2-fluorobenzamide 98%
4-Bromo-N-cyclopropyl-2-fluorobenzamide

Precaución: for Research Use Only

Formula: C₁₀H₉BrFNO
MDL Number: MFCD09943270
CAS Number: 877383-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR