1-Acetyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 97%

Supplier: Ambeed
Warning

A781654-100MG A781654-1G A781654-250MG
77006-424EA 33.95 USD
77006-424 77006-426 77006-428
1-Acetyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 97%
1-Acetyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MW: 188.18 g/mol
MDL Number: MFCD12027500
CAS Number: 155819-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR