5-(Aminomethyl)pyrrolidin-2-one hydrochloride 96%

Supplier: Ambeed
Warning

A781373-5G A781373-1G A781373-250MG
77006-422EA 239 USD
77006-422 77882-690 77882-691
5-(Aminomethyl)pyrrolidin-2-one hydrochloride 96%
5-(Aminomethyl)pyrrolidin-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁ClN₂O
MW: 150.61 g/mol
MDL Number: MFCD09701392
CAS Number: 115307-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR