1,3-Bis(2,6-diisopropylphenyl)imidazolidin-2-ylidene 98%

Supplier: Ambeed
Warning

A780250-250MG A780250-5G A780250-100MG
76993-188EA 50 USD
76993-188 76993-190 77886-748
1,3-Bis(2,6-diisopropylphenyl)imidazolidin-2-ylidene 98%
1,3-Bis(2,6-diisopropylphenyl)imidazolidin-2-ylidene

Precaución: for Research Use Only

Formula: C₂₇H₃₈N₂
MDL Number: MFCD10567379
CAS Number: 258278-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR