1-(3-Bromo-2-fluorophenyl)ethanol 98+%

Supplier: Ambeed
Warning

A779692-100MG A779692-1G A779692-250MG
76993-182EA 21.22 USD
76993-182 76993-184 76993-186
1-(3-Bromo-2-fluorophenyl)ethanol 98+%
1-(3-Bromo-2-fluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₈BrFO
MDL Number: MFCD16140247
CAS Number: 1221715-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR