Potassium 2,5-dihydroxybenzenesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dihydroxybenzenesulphonic acid potassium sal, Hydroquinonesulphonic acid potassium salt, Potassium 2,5-dihydroxybenzenesulphonate

A779308-100G A779308-25G A779308-500G A779308-5G
77006-400EA 21.22 USD
77006-400 77006-402 77006-404 77006-406
Potassium 2,5-dihydroxybenzenesulfonate 98%
Potassium-2,5-dihydroxybenzenesulphonate

Precaución: for Research Use Only

Formula: C₆H₅KO₅S
MW: 228.27 g/mol
MDL Number: MFCD00007475
CAS Number: 21799-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR