3-Bromo-5-phenoxypyridine 95%

Supplier: Ambeed
Warning

A777872-100MG A777872-1G
77036-294EA 24.41 USD
77036-294 77036-296
3-Bromo-5-phenoxypyridine 95%
3-Bromo-5-phenoxypyridine

Precaución: for Research Use Only

Formula: C₁₁H₈BrNO
MW: 250.09 g/mol
MDL Number: MFCD10698710
CAS Number: 28232-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR