2,3,4-Trifluoroanisole 95%

Supplier: Ambeed
Warning

A777447-250MG A777447-25G A777447-5G
77006-386EA 5 USD
77006-386 77006-388 77006-390
2,3,4-Trifluoroanisole 95%
2,3,4-Trifluoroanisole

Precaución: for Research Use Only

Formula: C₇H₅F₃O
MDL Number: MFCD00042396
CAS Number: 203245-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR