Methyl 1,4-Bis(Boc)-2-piperazinecarboxylate 95%

Supplier: Ambeed
Warning

A776565-250MG A776565-1G A776565-10G
76993-180EA 38.2 USD
76993-180 77573-494 77684-717
Methyl 1,4-Bis(Boc)-2-piperazinecarboxylate 95%
Methyl 1,4-Bis(Boc)-2-piperazinecarboxylate

Precaución: For research use only.

Formula: C₁₆H₂₈N₂O₆
MW: 344.4 g/mol
MDL Number: MFCD16621219
CAS Number: 171504-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR