4,4'-Dinonyl-2,2'-bithiazole 98+%

Supplier: Ambeed
Warning

A776544-100MG A776544-250MG
77019-210EA 61.54 USD
77019-210 77019-212
4,4'-Dinonyl-2,2'-bithiazole 98+%
4,4'-Dinonyl-2,2'-bithiazole

Precaución: for Research Use Only

Formula: C₂₄H₄₀N₂S₂
MW: 420.72 g/mol
MDL Number: MFCD27923057
CAS Number: 180729-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR