N-(3-Chloro-4-(10,11-dihydro-5H-benzo[e]pyrrolo[1,2-a][1,4]diazepine-10-carbonyl)phenyl)-5-fluoro-2-methylbenzamide 95%
Supplier: Ambeed
|
Warning
|
A776512-10MG
A776512-1MG
A776512-25MG
A776512-2MG
A776512-5MG
77006-366EA
68
USD
77006-366
77006-368
77006-370
77006-372
77006-374
N-(3-Chloro-4-(10,11-dihydro-5H-benzo[e]pyrrolo[1,2-a][1,4]diazepine-10-carbonyl)phenyl)-5-fluoro-2-methylbenzamide 95%
N-(3-Chloro-4-(10,11-dihydro-5H-benzo[e]pyrrolo[1,2-a][1,4]diazepine-10-carbonyl)phenyl)-5-fluoro-2-methylbenzamide
Precaución: for Research Use Only
Formula:
C₂₇H₂₁ClFN₃O₂ |
MDL Number:
MFCD00937905 CAS Number: 168079-32-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...