Methyl pent-4-ynoate 95%

Supplier: Ambeed
Danger

Synonyms: 4-Pentynoic acid methyl ester

A776415-1G A776415-5G A776415-25G
77019-208EA 16.98 USD
77019-208 77552-396 77573-490
Methyl pent-4-ynoate 95%
Methyl pent-4-ynoate

Precaución: for Research Use Only

Formula: C₆H₈O₂
MW: 112.127 g/mol
MDL Number: MFCD12025043
CAS Number: 21565-82-2
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR