5-(Boc-amino)-1,2,4-thiadiazole 98%

Supplier: Ambeed
Warning

A776247-1G A776247-5G
77019-204EA 8 USD
77019-204 77019-206
5-(Boc-amino)-1,2,4-thiadiazole 98%
5-(Boc-amino)-1,2,4-thiadiazole

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O₂S
MDL Number: MFCD18375242
CAS Number: 264600-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR