tert-Butyl 4-((4-(methoxycarbonyl)piperidin-1-yl)methyl)piperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A775212-1G A775212-250MG
77019-192EA 168.69 USD
77019-192 77019-194
tert-Butyl 4-((4-(methoxycarbonyl)piperidin-1-yl)methyl)piperidine-1-carboxylate 95%
tert-Butyl 4-((4-(methoxycarbonyl)piperidin-1-yl)methyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₃₂N₂O₄
MW: 340.45 g/mol
MDL Number: MFCD08275716
CAS Number: 914347-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR