1-(4-Bromophenyl)cyclobutane-1-carbonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Bromophenyl)cyclobutanecarbonitrile

A774458-250MG A774458-1G
77036-290EA 37.14 USD
77036-290 77602-982
1-(4-Bromophenyl)cyclobutane-1-carbonitrile 95%
1-(4-Bromophenyl)cyclobutane-1-carbonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrN
MW: 236.108 g/mol
MDL Number: MFCD10688823
CAS Number: 485828-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR