2,2'-Bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,1'-biimidazole 98%

Supplier: Ambeed
Warning

A771914-100G A771914-5G A771914-25G
77006-360EA 48 USD
77006-360 77006-362 77674-118
2,2'-Bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,1'-biimidazole 98%
2,2'-Bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,1'-biimidazole

Precaución: for Research Use Only

Formula: C₄₂H₂₈Cl₂N₄
MDL Number: MFCD00060081
CAS Number: 1707-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR