(2S,6S)-2,6-Dimethylpiperazine dihydrochloride 97%

Supplier: Ambeed
Warning

A771245-100MG A771245-1G A771245-250MG A771245-5G
76993-146EA 6 USD
76993-146 76993-148 76993-150 76993-152
(2S,6S)-2,6-Dimethylpiperazine dihydrochloride 97%
(2S,6S)-2,6-Dimethylpiperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆Cl₂N₂
MDL Number: MFCD11110543
CAS Number: 162240-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR