7-Benzyloxy-1H-indazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A771087-100MG A771087-250MG
77019-188EA 85.94 USD
77019-188 77019-190
7-Benzyloxy-1H-indazole-3-carboxylic acid 97%
7-Benzyloxy-1H-indazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂O₃
MW: 268.26 g/mol
MDL Number: MFCD05663992
CAS Number: 177941-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR