3-(Benzyloxy)-6-bromo-4-methoxypicolinic acid ≥95%

Supplier: Ambeed
Warning

A769831-100MG A769831-1G A769831-250MG
77019-182EA 163.38 USD
77019-182 77019-184 77019-186
3-(Benzyloxy)-6-bromo-4-methoxypicolinic acid ≥95%
3-(Benzyloxy)-6-bromo-4-methoxypicolinic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrNO₄
MW: 338.15 g/mol
MDL Number: MFCD27939567
CAS Number: 321596-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR