2-Methyl-5-(trifluoromethyl)phenylboronic acid 96%

Supplier: Ambeed
Warning

A769449-100MG A769449-1G A769449-250MG A769449-25G A769449-5G
77036-258EA 15.92 USD
77036-258 77036-260 77036-262 77036-264 77036-266
2-Methyl-5-(trifluoromethyl)phenylboronic acid 96%
2-Methyl-5-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₂
MW: 203.9541 g/mol
MDL Number: MFCD11042432
CAS Number: 947533-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR