Zafirlukast 99%

Supplier: Ambeed
Warning

Synonyms: N-[3-[[2-Methoxy-4-[[[(2-methylphenyl)sulphonyl]amino]carbonyl]phenyl]methyl]-1-methyl-1H-indol-5-yl]carbamic acid Cyclopentyl Ester, Cyclopentyl (3-(2-methoxy-4-((o-tolylsulphonyl)carbamoyl)benzyl)-1-methyl-1H-indol-5-yl)carbamate, Cyclopentyl N-[3-[[2-Methoxy-4-[[[(2-methylphenyl)sulphonyl]amino]carbonyl]phenyl]methyl]-1-methyl-1H-indol-5-yl]carbamate

A769198-10MG A769198-1MG A769198-25MG A769198-50MG
77006-348EA 24 USD
77006-348 77006-350 77006-352 77006-354
Zafirlukast 99%
Zafirlukast

Precaución: for Research Use Only

Formula: C₃₁H₃₃N₃O₆S
MW: 575.68 g/mol
MDL Number: MFCD00864775
CAS Number: 107753-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR