Methyl 3-chloro-2-methoxybenzoate 95+%

Supplier: Ambeed
Warning

A768606-1G A768606-250MG A768606-5G
77019-176EA 12 USD
77019-176 77019-178 77019-180
Methyl 3-chloro-2-methoxybenzoate 95+%
Methyl 3-chloro-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MDL Number: MFCD12547158
CAS Number: 92992-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR