Tetraphenylphosphonium iodide 98%

Supplier: Ambeed
Danger

A768351-25G A768351-5G
77036-246EA 30 USD
77036-246 77036-248
Tetraphenylphosphonium iodide 98%
Tetraphenylphosphonium iodide

Precaución: for Research Use Only

Formula: C₂₄H₂₀IP
MW: 466.3 g/mol
MDL Number: MFCD00011917
CAS Number: 2065-67-0
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR