4-Amino-1H-indazole-1-carboxylic acid tert-butyl ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-1H-indazole, N1-BOC protected, tert-Butyl 4-amino-1H-indazole-1-carboxylate

A768042-100MG A768042-1G A768042-250MG A768042-5G
77036-236EA 19 USD
77036-236 77036-238 77036-240 77036-242
4-Amino-1H-indazole-1-carboxylic acid tert-butyl ester 98%
4-Amino-1H-indazole-1-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃O₂
MDL Number: MFCD11052618
CAS Number: 801315-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR