2-Chloro-4'-fluoroacetophenone 98%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-1-(4-fluorophenyl)ethan-1-one, α-Chloro-4-fluoroacetophenone, 2-Chloro-1-(4-fluorophenyl)ethanone, Chloromethyl p-fluorophenyl ketone, 4-Fluorophenacyl chloride, 2-Chloro-4'-fluoroacetophenone

A767233-100G A767233-25G A767233-5G
77036-218EA 58.35 USD
77036-218 77036-220 77674-106
2-Chloro-4'-fluoroacetophenone 98%
2-Chloro-4'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆ClFO
MW: 172.59 g/mol
MDL Number: MFCD00011652
CAS Number: 456-04-2
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR