4,5-Dichloroimidazole 98%

Supplier: Ambeed
Warning

A766453-1G A766453-25G A766453-5G
77006-338EA 9 USD
77006-338 77006-340 77006-342
4,5-Dichloroimidazole 98%
4,5-Dichloroimidazole

Precaución: for Research Use Only

Formula: C₃H₂Cl₂N₂
MDL Number: MFCD00005195
CAS Number: 15965-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR