3-Bromo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A766118-100MG
A766118-1G
A766118-250MG
A766118-25MG
A766118-50MG
77019-160EA
337.37
USD
77019-160
77019-162
77019-164
77019-166
77019-168
3-Bromo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylic acid 95%
3-Bromo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₉BrO₂S MW: 261.13 g/mol |
MDL Number:
MFCD27996773 CAS Number: 188240-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...