Methyl 5-chloro-2,2-dimethylpentanoate 95%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A765612-250MG
A765612-1G
76993-128EA
39
USD
76993-128
77684-703
Methyl 5-chloro-2,2-dimethylpentanoate 95%, Ambeed.Inc
Methyl 5-chloro-2,2-dimethylpentanoate
Precaución: For research use only.
Formula:
C₈H₁₅ClO₂ MW: 178.65 g/mol |
MDL Number:
MFCD07782111 CAS Number: 73441-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...