(3,4-Dibromothiophen-2-yl)boronic acid 96%

Supplier: Ambeed
Danger

A765081-100MG A765081-1G A765081-250MG
77019-154EA 36.08 USD
77019-154 77019-156 77019-158
(3,4-Dibromothiophen-2-yl)boronic acid 96%
(3,4-Dibromothiophen-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₄H₃BBr₂O₂S
MW: 285.75 g/mol
MDL Number: MFCD07781231
CAS Number: 1256355-38-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR