4-(Chloromethyl)oxazole hydrochloride 95%

Supplier: Ambeed
Danger

A764919-100MG A764919-1G A764919-250MG
77006-332EA 137 USD
77006-332 77006-334 77006-336
4-(Chloromethyl)oxazole hydrochloride 95%
4-(Chloromethyl)oxazole hydrochloride

Precaución: for Research Use Only

Formula: C₄H₅Cl₂NO
MW: 153.99 g/mol
MDL Number: MFCD18205958
CAS Number: 675149-75-4
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR