Benzyl (2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 95%

Supplier: Ambeed
Warning

A764272-1G A764272-250MG
77019-146EA 62.6 USD
77019-146 77019-148
Benzyl (2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 95%
Benzyl (2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₂₀H₂₃BFNO₄
MW: 371.21 g/mol
MDL Number: MFCD17015818
CAS Number: 1256359-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR