(3-((4-(2-Methoxyethyl)phenoxy)methyl)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A763595-1G A763595-250MG A763595-5G
77019-140EA 38.2 USD
77019-140 77019-142 77019-144
(3-((4-(2-Methoxyethyl)phenoxy)methyl)phenyl)boronic acid 95%
(3-((4-(2-Methoxyethyl)phenoxy)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₉BO₄
MW: 286.13 g/mol
MDL Number: MFCD07784393
CAS Number: 871126-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR