4-Chloro-5-nitro-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A762791-100MG A762791-1G A762791-250MG A762791-5G
77036-186EA 14.86 USD
77036-186 77036-188 77036-190 77036-192
4-Chloro-5-nitro-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine 95%
4-Chloro-5-nitro-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₁₃H₈ClN₃O₄S
MW: 337.73 g/mol
MDL Number: MFCD18072770
CAS Number: 1245649-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR