2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 2,2,2-Trifluoro-1-(pyridin-3-yl)ethanamine hydrochloride

A762307-100MG A762307-1G A762307-250MG
77006-320EA 48.81 USD
77006-320 77006-322 77006-324
2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride 95%
2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₇F₃N₂·HCl
MW: 212.60 g/mol
MDL Number: MFCD06739079
CAS Number: 1138011-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR